Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Buckets for 4-Place Swinging Bucket Rotors
Round or rectangular buckets for use with Four-Place Swinging Bucket Rotors 75006445 and 75006434
472.35 € - 550.00 €
Specifications
For Use With (Equipment) Swinging bucket rotor
View More Specs
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10458191
View accessory products View Documents
Thermo Scientific™
75006441
Round Bucket; Capacity: 1 x 750 mL
472.35 €
Each
10638952
View Documents
Thermo Scientific™
75006446
Rectangular Bucket, 1 x 250 mL
550.00 €
Each
Description
Description
Order Info
Sold individually; order correct quantity to equip rotor. Sealing lids sold separately. Accessory adapters accept assorted tube styles and volumes from 1.5 to 250mL.
Specifications
Specifications
Swinging bucket rotor