missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph™ SP Quick Single-channel Pump Heads
Fast and convenient tube changes
Brand: Heidolph™ 527-11300-00
536.75 EUR valid until 2024-11-30
Use promo code "21681" to get your promotional price.
This item is not returnable.
View return policy
Description
- Low pulsation due to 5 roller system
- Pump head features ball bearings
- Rollers made of stainless steel
- Depending on drive and tubing, flow rate ranges from 0.38 to 3436 mL/min.
Specifications
0.38 to 3436 mL/min. | |
Single-channel pumps; tubing (wt 2.5 mm) | |
Pump Head |