missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Frigimat™ Junior Dry Ice Maker

Produces dry ice in flake or snow form

Brand:  Bel Art™ F38886-0000

 View more versions of this product

Product Code. 12952580

  • 287.00 € / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket

Includes:

Brass nozzle with polypropylene cone, clamp, heavy-duty cloth bag

This item is not returnable. View return policy

Description

Description

  • Screw nozzle directly to cylinder of compressed LCO2; cylinder must have a dip tube
  • When the cylinder valve is opened about 0.75 turn, LCO2 expands into the bag to create dry ice snow; The valve is then closed and the bag is ready to be removed and emptied
  • Secure bag to cone on the nozzle with the clamp
  • After emptying, the bag can be immediately reattached for next filling
  • Makes .77 kg (1.7 lb) of flakes per bag

Order Info

Cone, 5L x 2.312 in. dia. (127 x 59mm) at widest end. Bag, 6 dia. x 12 in. L (15 x 31cm).

Specifications

Specifications

0.77kg
Standard CO2 cylinder
Brass, Polypropylene
Dry Ice Maker
Making Dry Ice
Brass nozzle with Polypropylene cone, clamp, heavy-duty cloth bag
Junior Dry Ice Maker
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel Art™ Frigimat™ Junior Dry Ice Maker > Easy and fast dry ice flake/snow production

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.