missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Heavy-Duty Test-Tube Racks

These 3 tier heavy-duty test tube racks have hand grips in the end plates for easy transport

Brand:  Bel-Art™ F18856-0000

 View more versions of this product

Product Code. 10728151

  • 44.75 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stackable, compact and steam autoclavable at 121°C (250°F) for durability in the lab
  • End plates have hand grips for easy transport
  • Base tier has rounded wells with drain holes, and the top tier has lettered and numbered rows
  • Available in different sizes
  • 8.3 x 4.3 in. (21.3 x 11cm)

Specifications

Specifications

Test Tube Rack
Tubes (13 to 16 mm)
50
8.37 x 4.31 x 4.37 in. (21.3 x 11 x 11.1cm)
8.37 in.
4.31 in.
4.37 in.
Autoclavable
Polypropylene
Heavy-duty test tube
21.3 cm
11 cm
11.1 cm
16 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.