missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ LabMat™ Bench Liners

Simplify cleanup and safeguard work surfaces from stains, spills and wear. Bel Art™ SP Scienceware™ LabMat™ Bench Liners can be rolled up and discard after contamination.

Brand:  Bel-Art™ F24675-0020

 View more versions of this product

Product Code. 12918837

  • 273.65 € / Pack of 100
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
  • Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
  • Cut to size easily with scissors or the Scienceware™ Safety Cutter (Mfr. No. 21012-0000)
  • Roll up and discard in proper waste container after contamination
  • Select orange to identify use with biohazard materials
Specifications

Specifications

Bench Protector
400 mL/m2
610 mm
508 mm
White
Single-use
6 Case
Polyethylene
24 in.
20 in.
24 x 20 in. (610 x 508mm)
Pack of 100 sheets
Simplify cleanup and safeguard work surfaces from stains, spills and wear
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers