missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Polypropylene Sterilizing Tray Covers
Covers for use with SP Scienceware™ Polypropylene Sterilizing Trays.
Brand: Bel-Art™ H16263-0000
Description
- Corrosion-proof polypropylene
- Rounded corners for easy cleaning and smooth, tapered sides for stacking.
- Covers are 3.2mm (1/8 in.) thick with a 25.4mm (1 in.) skirt that fits snugly over the rim of the tray
- Steam autoclavable at 121°C (250°F)
Specifications
18 L | |
2.54 cm | |
Polypropylene | |
39.4 cm | |
20.3 in. | |
Rectangular | |
15.5 in. |
20.3 x 15.5 x 1 in. (53.3 x 39.4 x 2.54 cm) | |
53.3 cm | |
Tray Cover | |
Cover fits 18L Sterilizing Tray, Mfr. No. H162640000 | |
1 in. | |
3.2 mm | |
Autoclavable |