Learn More
Bel-Art™ SP Scienceware™ Sterile Economy Sample Spoons
For retrieving small, specific volume of powdered or liquid samples
Brand: Bel-Art™ F36740-0410
This item is not returnable.
View return policy
Description
- Made from high-impact polystyrene, FDA grade and sterilized by gamma irradiation
- Comes with flat bottom shape to allow use as weighing boat
- Available in white color
- Available in individual packs for one time use
- Includes certificate of processing for sterilization
Specifications
Spoon | |
Certificate of processing for sterilization | |
8 mL | |
Sterile | |
FDA |
Polystyrene | |
Flat | |
Single-use | |
White | |
Ideal for retrieving small, specific volume samples of powders or liquids, provides durability and stiffness when sampling packed powders |