missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Sterile Eye Wash Solution Stations
Provide effective and immediate eye irrigation by outfitting your workspace with the SP Scienceware™ Sterile Eye Wash Solution Station.
28.36 €
Specifications
Capacity (English) | 33.81 oz. |
---|---|
Capacity (Metric) | 1000 mL |
Sterility | Sterile |
Product Code | Brand | Color | Height (English) | Width (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Height (English) | Width (English) | Price | Quantity & Availability | |||
10286354
|
Bel-Art™
F24879-0032 |
Natural | 7.5 in. | 3.56 in. | |||||
Description
- Choose from single or double stations with 1000mL (32 oz.) bottles of sterile, isotonic, buffered eye wash solution, ready for immediate use
- Safety-green eye wash stations can be wall-mounted for quick access when an emergency occurs
- Include quick instruction diagrams, multi-language instructions, a polished metal mirror and four holes for mounting with screws (hardware not included)
- Bottles have twist-off caps and a three year shelf life from date of manufacture
- Sterile irrigation solution is helpful in rinsing away chemical contaminants and other harmful irratants and is similar in pH value to that of human tears
Specifications
33.81 oz. | |
1000 mL | |
Sterile |