missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Stopcocks

With a removable, leak-proof plug made of chemical-resistant, non-stick PTFE with barbed tubulations that accept 1/4 to 3/8 in. I.D. tubing

Brand:  Bel-Art™ F30890-0000

 View more versions of this product

Product Code. 10018220

  • 69.92 € / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Instant cut-off control
  • Pressure rating is 240kPa (35psi)
  • Steam autoclavable at 121°C (250°F)
  • Three-way model allows connection of any two or all three arms
Specifications

Specifications

2-way Stopcock
2-way; 2mm bore size
Autoclavable
6.4 to 9.5 mm
Removable Leak-Proof Plug
2 mm
Polypropylene, PTFE
240 KPa
1 Case
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.