missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Super Polyfoil™ Flask Wrap

Provides a chemically inert barrier to cover vessels for autoclaving, incubating or storage. Bel Art™ SP Scienceware™ Super Polyfoil™ Flask Wrap folds easily and flattens without kinking.

Brand:  Bel-Art™ F13200-0088

 View more versions of this product

Product Code. 12339558

  • 65.90 € / 100 sheets
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

 

  • Hermoplastic layer is laminated to 1mL thick foil
  • Folds easily; flattens without kinking
  • Resistant to punctures, tears and corrosion
  • Polyester film blocks pinholes and resists organic solvents and caustic agents
  • Complies with FDA regulations governing direct food contact applications
  • Comes in a pack of 100

 

Specifications

Specifications

Clinical, research and industrial uses
Foil
203 mm
1 mil
FDA
203 mm
Aluminum, Polyester
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.