missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Tape for Protective Labeling Tape System

Tape for use with Bel Art Scienceware Protective Labeling Tape System (cat. no. 1186750)

Brand:  Bel-Art™ F13491-0150

60.92 EUR valid until 2025-02-13
Use promo code "24130" to get your promotional price.


 View more versions of this product

Product Code. 13425256

  • 64.20 € / Pack of 2
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The clear tape protects labels from the freezer, autoclave, acids, alkalis and organic solvents. Test results prove the tape remains securely affixed with no discoloration after being autoclaved for 40 minutes at 125°C (258°F), as well as after one week in liquid nitrogen at -196°C (-321°F). All tape sizes are 1mil (0.001 in.) thick with 1mil (0.001 in.) of acrylic adhesive.

  • Clear, polypropylene tape has high resistance to chemicals and moisture; provides great versatility for heavy-duty lab use

Specifications

Specifications

Autoclavable
Clear
Polypropylene
Heavy duty lab use
1 mil
32.9 m
Acrylic
Tape
Protective Lab Labeling Tape
2/Pk.
36 yd.
1.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.