missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Two-Color Wash Bottles - Safety-Vented™ and Safety Labeled, Wide-Mouth
Click to view available options
Capacity (Metric):
500 mL
Chemical Label:
Custom Write-on
Ethanol
Isopropanol
Methyl Ethyl Ketone
Description
A unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know Safety-Labeled Wash Bottles
Volatiles are vented eliminating solvent drips. They are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.
- Valve eliminates drips; allows dispensing of wash bottle in an upright or inverted position
- Wide mouth 53mm (2 in.) bottle for easy refilling
- Polypropylene closure with leakproof design; color-coded to aid in easy identification
- Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
- Labeled with six of the most commonly used solvent names or label your own; custom labeling available
Order Info
8 oz. (250mL)
Specifications
Capacity | 16 oz., 500mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
Diameter (Metric) Outer Neck | 53mm |
For Use With (Application) | Ethanol |
Includes | Natural Polypropylene Cap |
Material | Polyethylene |
Quantity | 1Each |
Closure Color | Natural |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'