Learn More
Heidolph Instruments SP VARIO™ Single-channel Pump Head
Suitable for most Heidolph tubing due to adjustable roller spacing
Brand: Heidolph Instruments 523-45110-00
927.67 EUR valid until 2024-11-30
Use promo code "21681" to get your promotional price.
This item is not returnable.
View return policy
Description
- Convex rollers do not squeeze or crush organic cell cultures
- Pump head features ball bearings
- Rollers made of stainless steel and aluminum
- Depending on drive and tubing, the flow rate ranges from 2 to 4151 mL/min.
Specifications
LED | |
2.0 to 4151 mL/min. | |
IP 30 |