missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals SP600125, 98%, Thermo Scientific Chemicals Product Code.: 15498689

Thermo Scientific Chemicals SP600125, 98%, Thermo Scientific Chemicals

Product Code. 15498689
50 mg
Click to view available options
Quantity:
50 mg
100 mg
This item is not returnable. View return policy

Product Code. 15498689

Brand: Thermo Scientific Chemicals J67272.MF

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SP 600125 is a potent and reversible inhibitor of JNK1-3 with an IC50 value of 0.11 M.1 It is cell-permeable and dose-dependently inhibits c-Jun phosphorylation in cells, blocking the expression of COX-2 and TNF-α in monocytes and IL-10, TNF-α, and IFN-γ in T-cells. SP 600125 also prevents apoptosis in many cell types, including B-cells, and inhibits autophagy in HeLa cells. Competitively and reversibly inhibits JNK1, 2 and 3 (IC50 = 40 - 90 nM) with negligible activity at ERK2, p38β and a range of enzymes (IC50 > 10 ?M). Shown to have reduced selectivity over other protein kinases under certain conditions. Prevents BMP9-induced osteogenic differentiation of MSCs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-56-6
Molecular Formula C14H8N2O
Synonym 1,9-Pyrazoloanthrone;

Specifications

Quantity 50 mg
Description Yellow to brown
Content And Storage -30°C to -10°C
Formula Weight 220.23
Solubility Soluble in DMSO at approx. 20mg/ml
Product Type SP600125, 99%
Form Solid
InChI Key ACPOUJIDANTYHO-UHFFFAOYSA-N
SMILES O=C1C2=C3C(NN=C3C3=CC=CC=C13)=CC=C2
IUPAC Name 14,15-diazatetracyclo[7.6.1.0²,⁷.0¹³,¹⁶]hexadeca-1(15),2,4,6,9(16),10,12-heptaen-8-one
Molecular Weight (g/mol) 220.23
Assay 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals SP600125, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.