Learn More
BRAND™ Spare Lateral Stopcock with Key
Stopcock housing with burette tip includes interchangeable PTFE or borosilicate glass key with retention device.
20.35 € - 42.75 €
Specifications
Capacity (Metric) 50 mL
For Use With (Equipment) Burette
No. per Pack 1 Each
Includes Retention Device
Type Stopcock Key
View More Specs
Products 2
Product Code Brand Material Price Quantity & Availability  
Product Code Brand Material Price Quantity & Availability  
10278401
View Documents Promotion Details
BRAND™
81167
PTFE
42.75 €
Each
In Stock
to see stock.
Add to basket
10750043
View Documents Promotion Details
BRAND™
81142
Borosilicate Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Lateral stopcock housing with burette tip
  • Interchangeable PTFE or borosilicate glass key with retention device
Specifications
Specifications
50 mL
1 Each
Stopcock Key
Burette
Retention Device