missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liebherr™ Spark-Free Upright Lab Freezer with Shelves

Secure, accurate and ultra energy-efficient, spark-free interior, freezer storage, the Liebherr™ Spark-Free Interior, Lab Freezer is designed for general purpose applications with Liebherr build-quality you can rely on
Brand: Liebherr™ SFFFG4001
2679.00 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
This item is not returnable.
View return policy
Description
- Spark-Free upright Lab Freezer for storage of solvents and reagents, reach-in, 1-section
- Temperature range -9 to -30°C, ± 5°C temperature stability tested to EN60068-3
- White exterior, commercial-grade molded polystyrol interior, static refrigeration with manual defrost
- Refrigerant R 290, connection rating 2.0 A, 220-240V/50/1-ph
- This model is intended for the spark-free storage
- ATEX-certified spark-free interior
- Multifunctional digital controller with high resolution display and connectivity options
- Door lock, reversible hinges, antimicrobial lever handle
- Accessories: optional set of drawers available
Specifications
Atex Freezer | |
-10°C to -30°C | |
-20°C | |
Reversible, Self-closing, Fitted Lock | |
Scratch-resistant Painted Steel | |
Commercial-grade Molded Polystyrol | |
4 | |
Upright | |
Optional (80 to 100 mm dia.) | |
Low-maintenance Rear-wall Wire-on-tube | |
1 x 10 mm dia. | |
Manual | |
SmartCoolingHub | |
CEE 7/7 | |
1, Single Phase | |
384 kWh/a | |
R 290 |
316 L | |
NTC | |
±5 K | |
1 | |
Stainless Steel | |
White Powder-coated | |
ISO 9001, ISO 14001, CE | |
Touch Control, Digital Display in Door | |
White | |
Storage of Temperature-sensitive Products | |
2 yr. Parts and Labor | |
SFFfg 4001 | |
2.0 A | |
220/240 V | |
120 W | |
CFC-free |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction