missing translation for 'onlineSavingsMsg'
Learn More

Liebherr™ Spark-Free Upright Lab Freezer with Shelves
GREENER_CHOICE

Product Code. p-200006736
Click to view available options
Capacity (Metric):
316 L
499 L
Unit Size:
Each

Product Code. 17869106

Brand: Liebherr™ SFFFG4001

Please to purchase this item. Need a web account? Register with us today!

Secure, accurate and ultra energy-efficient, spark-free interior, freezer storage, the Liebherr™ Spark-Free Interior, Lab Freezer is designed for general purpose applications with Liebherr build-quality you can rely on

  • Spark-Free upright Lab Freezer for storage of solvents and reagents, reach-in, 1-section
  • Temperature range -9 to -30°C, ± 5°C temperature stability tested to EN60068-3
  • White exterior, commercial-grade molded polystyrol interior, static refrigeration with manual defrost
  • Refrigerant R 290, connection rating 2.0 A, 220-240V/50/1-ph
  • This model is intended for the spark-free storage
  • ATEX-certified spark-free interior
  • Multifunctional digital controller with high resolution display and connectivity options
  • Door lock, reversible hinges, antimicrobial lever handle
  • Accessories: optional set of drawers available 

Specifications

Type Atex Freezer
Capacity (Metric) 316 L
Temperature Range -10°C to -30°C
Temperature Sensor NTC
Temperature (Setpoint) -20°C
Temperature Stability ±5 K
Door Style Reversible, Self-closing, Fitted Lock
No. of Doors 1
Cabinet Material Scratch-resistant Painted Steel
Chamber Material Stainless Steel
Interior Commercial-grade Molded Polystyrol
Exterior Finish White Powder-coated
Shelves 4
Certifications/Compliance ISO 9001, ISO 14001, CE
Style Upright
Display Touch Control, Digital Display in Door
Casters Optional (80 to 100 mm dia.)
Color White
Condenser Type Low-maintenance Rear-wall Wire-on-tube
For Use With (Application) Storage of Temperature-sensitive Products
Ports 1 x 10 mm dia.
Warranty 2 yr. Parts and Labor
Defrost Manual
Model SFFfg 4001
Monitoring Options SmartCoolingHub
Amperage 2.0 A
Plug Type CEE 7/7
Voltage 220/240 V
Phase 1, Single Phase
Wattage 120 W
Energy Usage 384 kWh/a
Compressor CFC-free
Refrigerant R 290
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.