missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liebherr™ Spark-Free Upright Lab Fridge with Shelves
Secure, accurate and ultra energy-efficient, spark-free interior, fridge storage, the Liebherr™ Spark-Free Interior, Lab Fridge is designed for general purpose applications with Liebherr build-quality you can rely on.
Brand: Liebherr™ SRFfg 3501
2232.50 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- Spark-Free upright Lab Fridge for storage of solvents and reagents, reach-in, 1-section
- Temperature range +3 to +16°C, +/- 3°C temperature stability tested to EN60068-3
- white exterior, commercial-grade molded polystyrol interior, fan-assisted refrigeration
- Refrigerant R 600a, connection rating 2.0 A, 220-240V/50/1-ph
- This model is intended for the spark-free storage
- ATEX-certified spark-free interior
- Multifunctional digital controller with high resolution display and connectivity options
- Door lock, reversible hinges, antimicrobial lever handle
Specifications
Atex Fridge | |
3°C to 16°C | |
+5°C | |
R 600a | |
1 | |
Scratch-resistant Painted Steel | |
Commercial-grade Molded Polystyrol | |
Upright | |
Optional (80 to 100 mm dia.) | |
Low-maintenance Rear-wall Wire-on-tube | |
1 x 10 mm dia. | |
2 yr. Parts and Labor | |
SRFfg 3501 | |
Yes | |
2.0 A | |
220/240 V | |
406 kWh/a |
344 L | |
NTC | |
±3 K | |
Solid Reversible Self-closing with Lock | |
4 | |
White Powder-coated | |
ISO 9001, ISO 14001, CE | |
Touch Control, Digital Display in Door | |
White | |
Storage of Temperature-sensitive Products | |
Scientific Performance | |
Timed Off-cycle | |
SmartCoolingHub | |
CFC-free | |
CEE 7/7 | |
120 W |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction