Learn More
Labcold™ Sparkfree Freezers
For the safe storage of potentially flammable items. Labcold™ Sparkfree Freezers come in both chest and upright models.
Brand:  Labcold™ RLCF0720
 View more versions of this product
Product Code. 11528954
  • 3970.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CFC and HCFC free
  • Operator controlled defrost
  • Alarm/temperature display can be fitted as an optional extra
  • Chest freezers feature integral lid lock
  • Locks available for all upright freezers as an additional extra
Specifications
Specifications
215 L
1 Basket
RLCF0720
860 mm
-18°C to -25°C
Chest
730 mm
725 mm