Learn More
3M™ Aura™ Specialty Respirator, Series 9300
Lightweight and foldable. The 3M Aura Particulate Respirator uses a unique three-panel design for greater comfort and easy communication.
87.00 € - 2195.00 €
Product Code | Brand | Filter Efficiency | With Exhalation Valve | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Efficiency | With Exhalation Valve | Color | Price | Quantity & Availability | |||
12352399
|
3M™
7000034739 |
FFP1 | Yes | Gray |
87.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||
11763373
|
3M™
7000034737 |
FFP1 | No | Blue, Black | N/A | ||||
11797564
|
3M™
7000034738 |
FFP2 | Yes | Blue |
121.00 €
Pack of 10 |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||
15263059
|
3M™
7000052909 |
FFP3 | Yes | White |
2195.00 €
Pack of 180 |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||
12352389
|
3M™
7000034740 |
FFP1 | No | Gray |
142.00 €
Pack of 20 |
In Stock
Log in to see stock. |
|||
Description
Fits a wide range of face shapes and sizes- Low breathing resistance filter technology for easier breathing
- Ingenious three-panel design accommodates greater facial movement during speech for more comfortable wear
- An embossed top panel to reduce fogging of eyewear
- A sculpted nose panel for improved compatibility with eyewear
- An innovative chin tab for ease of donning and adjustment of the respirator on the face
- Supplied in hygienic packaging to help protect the respirator from contamination before use, also allows practical storage and dispensing in the workplace.
- Cool Flow™ valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions