missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Specimen Bag with Document Wallet, Plain

Specimen bag ideal for safe patient sample transport.

Brand:  Wheaton™ 370160

113.05 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 16404989

  • 122.00 € / Pack of 1000
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ensures safe sample transfer
  • For transportation of clinical specimens and documents
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Specimen Bag
Self-adhesive
Transportation of clinical specimens and documents
LDPE
1000
150 x 150 mm
Not Autoclavable
Single-use
Document Pouch, Plain
Non-sterile
100
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers