Learn More
Spectrum Labs™ Spectra/Por™ 1 Membrane Disc
Regenerated cellulose membrance for use with Spectrum Equilibrium Dializer and Spectra/Por MacroDializer systems
Brand:  Spectrum Labs™ 132474
 View more versions of this product
Product Code. 12717526
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
For use with Spectrum Equilibrium Dialyzer and Spectra/Por™ MacroDialyzer systems.
Regenerated cellulose (RC) precut membrane discs
Supplied dry with a glycerol as a humectant
Specifications
Specifications
Membrane Disc
Regenerated Cellulose
For Basic Dialysis
6 to 8 kDa
100 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Spectrum Labs™ Spectra/Por™ 1 Membrane Disc >