missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Spectra/Por™ 1-4 Standard RC Dialysis Membrane in Flat Sheets
Clear, flexible and sturdy Standard RC (regenerated cellulose) in single layer square sheets that can be used with a variety of dialysis equipment or can be cut into specific dimensions for attaching to support frames for customized diagnostic assays, equilibrium studies, desalting, protein purification, etc.
Brand: Spectrum™ 132677
531.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Flat-Sheets deal for use with dialysis equipment or cutting to size
- Available in 3.5 kD, 6-8 kD and 12-14 kD MWCO's
- Good chemical compatibility with dilute strong acids & bases, conc. weak acids and bases, many alcohols & some organics.
- Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
- 25 sheets/pkg
Do not tie knots in tubing; use closures
Specifications
Spectra/Por 1 Dialysis Membrane Flat Sheets | |
Spectra/Por 1 | |
240 mm | |
240 mm | |
Clear | |
4°C to 121°C | |
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting |
6 to 8 kDa | |
9.4 in. | |
9.4 in. | |
9.4 x 9.4 in. (240 x 240 mm) | |
39°F to 250°F | |
25 Sheets |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction