missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ 6 Pre-wetted Standard RC Dialysis Tubing, 1-50 kD MWCO

Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane

Brand:  Spectrum™ 132550

450.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11425899

  • 462.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane

This ready-to-use membrane is packaged wet in a preservative solution of 0.05% sodium azide. Simply rinse away the preservative solution with DI water, load the sample and dialyze.

  • Pre-wetted & ready-to-use, no glycerin removal required
  • 8 MWCO's ranging from 1 kD - 50 kD
  • Good chemical compatibility with dilute strong acids and bases, conc. weak acids & bases, many alcohols and some organics
  • Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
  • Ideal for sample prep, desalting, buffer exchange, pH change, protein purification, etc.
  • Economic 10 meters/pkg

Do not tie knots in tubing; use closures

Specifications

Specifications

Spectra/Por 6 Dialysis Tubing
Regenerated Cellulose
0.29 in.
10 m
39.2°F to 249.8°F
Each
25 kDa
7.5 mm
33 ft.
Packaged wet in 0.05% sodium azide
4°C to 121°C
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.