Learn More
Spectrum™ Spectra/Por™ 6 Pre-wetted Standard RC Dialysis Tubing, 1-50 kD MWCO
Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane
Brand: Spectrum™ 132540
450.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane
This ready-to-use membrane is packaged wet in a preservative solution of 0.05% sodium azide. Simply rinse away the preservative solution with DI water, load the sample and dialyze.
- Pre-wetted & ready-to-use, no glycerin removal required
- 8 MWCO's ranging from 1 kD - 50 kD
- Good chemical compatibility with dilute strong acids and bases, conc. weak acids & bases, many alcohols and some organics
- Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
- Ideal for sample prep, desalting, buffer exchange, pH change, protein purification, etc.
- Economic 10 meters/pkg
Do not tie knots in tubing; use closures
Specifications
Spectra/Por 6 Dialysis Tubing | |
Regenerated Cellulose | |
0.29 in. | |
10 m | |
39.2°F to 249.8°F | |
Each |
50 kDa | |
7.5 mm | |
33 ft. | |
Packaged wet in 0.05% sodium azide | |
4°C to 121°C | |
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.