Learn More
Spectrum™ Spectra/Por Biotech-Grade RC Dialysis Membrane Tubing
A premium dialysis membrane that provides superior molecular selectivity than Standard RC and superior tolerance than Biotech CE
Brand:  Spectrum™ 133342
 View more versions of this product
Product Code. 13542040
  • 481.00 € / Each
Explore more special offers
Description
Description
  • Crafted from a regeneration process that yields superior temperature tolerances and chemical compatibilities with the same high purity and selectivity as Biotech CE
  • Membrane can tolerate concentrated-weak acids and bases, dilute-strong acids and bases, most alcohols and some mild or dilute organic solvents
  • Rigidly controlled porosity for better selectivity and separations
  • Ultrapure, does not interfere with molecular activity and function
  • 3 MWCOs and 2 tubing flat-widths
  • Packaged dry with glycerin (soak 15 min. in water)
  • Use with pH 2 to 12 and temp 4 to 60°C
Specifications
Specifications
Spectra/Por Dialysis Tubing
Regenerated Cellulose
0.39 in.
5 m
39.2°F to 140°F
Each
20 kDa
10 mm
16.4 ft.
Packaged dry with glycerin (soak 15 min. in water)
4°C to 60°C
Frequent dialysis applications, desalting, purification, sample preparation