Learn More
Spectrum™ Labs 3.5 - 5 kD Biotech RC Dialysis Kits
With tubing made from Biotech RC membrane that can tolerate concentrated-weak acids, bases, dilute-strong acids, bases, most alcohols and some mild or dilute organic solvents
Brand:  Spectrum™ 133 192T
Product Code. 15350782
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Includes:
  • 0.5m Biotech RC tubing with glycerine
  • Two universal tubing closures
  • Five opening picks
Specifications
3.5 to 5 kDa
6.4 mm
0.32 mL/cm vol./length
0.5 m Tubing/Kit