Learn More
Spectrum™ Spectra/Mesh Woven Polymer Macrofiltration Squares: N (Nylon)
Combines high tensile strength with flexibility
Brand:  Spectrum™ 146486
 View more versions of this product
Product Code. 15370912
  • 414.00 € / Pack of 3
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Combines high tensile strength with flexibility
Ideal for straining, filtering and screening solutions or suspensions containing high-molecular-weight molecules or particles.
  • Nonabsorbent
  • Combines high tensile strength with flexibility
Specifications
Specifications
High
Nylon
Hydrophilic
3 to 10
180°C
Spectra Mesh Woven Filter
ISO 9001:2008
Sterile
50%
300μm
200 μm
4.65 in.2