Learn More
Spectrum™ Spectra/Por™ 7 Membrane Tubing (Sulfur and Heavy Metal Free):1 1000 to 50,000 Dalton MWCO
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
315.18 € - 631.00 €
Specifications
Product Type Spectra/Por 7 Membrane Tubing
Material Regenerated Cellulose
Length (English) 16.4 ft.
Length (Metric) 5 m
Color Transparent
View More Specs
Products 28
Product Code Brand Diameter (Metric) Quantity MWCO Price Quantity & Availability  
Product Code Brand Diameter (Metric) Quantity MWCO Price Quantity & Availability  
11415879
View accessory products View Documents
Spectrum™
132104
24 mm Each 1 kDa
631.00 €
Each
11425879
View accessory products View Documents
Spectrum™
132105
45 mm Each 1 kDa
631.00 €
Each
11435909
View accessory products View Documents
Spectrum™
132108
24 mm Each 2 kDa
615.00 €
Each
11445909
View accessory products View Documents
Spectrum™
132109
29 mm Each 2 kDa
626.00 €
Each
11425909
View accessory products View Documents
Spectrum™
132107
11.5 mm Each 2 kDa
626.00 €
Each
11455909
View accessory products View Documents
Spectrum™
132110
11.5 mm Each 3.5 kDa
626.00 €
Each
11475909
View Documents
Spectrum™
132112
34.4 mm Each 3.5 kDa
631.00 €
Each
11465909
View accessory products View Documents
Spectrum™
132111
29 mm Each 3.5 kDa
440.00 €
Each
11405919
View accessory products View Documents
Spectrum™
132113
7.5 mm Each 8 kDa
498.00 €
Each
11485909
View Documents
Spectrum™
132114
15 mm Each 8 kDa
580.00 €
Each
11425919
View Documents
Spectrum™
132131
32 mm Each 8 kDa
564.00 €
Each
11415919
View Documents
Spectrum™
132115
20.4 mm Each 8 kDa
403.00 €
Each
11495909
View Documents
Spectrum™
132116
25.5 mm Each 8 kDa
554.00 €
Each
11455919
View accessory products View Documents
Spectrum™
132117
7.5 mm Each 10 kDa
431.00 €
Each
11475919
View accessory products View Documents
Spectrum™
132119
20.4 mm Each 10 kDa
631.00 €
Each
11485919
View Documents
Spectrum™
132120
29 mm Each 10 kDa
615.00 €
Each
11425929
View accessory products View Documents
Spectrum™
132121
7.5 mm Each 15 kDa
391.00 €
Each
11445929
View accessory products View Documents
Spectrum™
132122
15 mm Each 15 kDa
391.00 €
Each
11455949
View accessory products View Documents
Spectrum™
132125
7.5 mm Each 25 kDa
503.00 €
Each
11435949
View accessory products View Documents
Spectrum™
132126
17.8 mm Each 25 kDa
575.00 €
Each
11465929
View accessory products View Documents
Spectrum™
132124
29 mm Each 15 kDa
554.00 €
Each
11445949
View Documents
Spectrum™
132127
21.6 mm Each 25 kDa
621.00 €
Each
11455929
View accessory products View Documents
Spectrum™
132123
20.4 mm Each 15 kDa
402.00 €
Each
10749445
View accessory products View Documents
Spectrum™
132128
7.5 mm Each 50 kDa
621.00 €
Each
10071954
View accessory products View Documents
Spectrum™
132129
17.8 mm Each 50 kDa
621.00 €
Each
10031954
View Documents
Spectrum™
132130
21.6 mm Each 50 kDa
631.00 €
Each
11435929
Zubehör anzeigen missing translation for 'viewDocuments'
Spectrum™
128458
3.8 mm 1 Pk. 15 kDa This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11465919
Zubehör anzeigen missing translation for 'viewDocuments'
Spectrum™
132118
15 mm Each 10 kDa
570.00 €
Each
Beschreibung
Beschreibung
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.
  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus
Do not tie knots in tubing; use closures
Spezifikation
Spezifikation
Spectra/Por 7 Membrane Tubing
16.4 ft.
Transparent
39.2°F to 249.8°F
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Regenerated Cellulose
5 m
Packaged wet in 0.05% sodium azide
4°C to 121°C
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt