Learn More
Spectrum™ Spectra/Por™ Biotech Cellulose Ester (CE) Dialysis Membrane Tubing
Selected MWCOs from 100 to 100,000 Daltons
Brand:  Spectrum™ 131342
 View more versions of this product
Product Code. 11405959
  • 685.00 € / Each
Explore more special offers
Description
Description
Selected MWCOs from 100 to 100,000 Daltons
  • CE tubing membranes are ideal for desalting, purification, and molecular separation
  • Supplied in 10m (33 ft.) lengths in two rolls of 5m each, in 0.1% sodium azide solution
Order Info
Note:Do not tie—must be used with Spectra/Por nylon closures (08-671-52, -53, -55). Strong polar solvents such as acetone, MEK, or dioxane will irreparably damage membranes. Contact Spectrum Technical Service, 1-800-634-3300, for more information.
Specifications
Specifications
Spectra/Por Dialysis Membrane Tubing
Cellulose Ester
0.39 in.
10 m
39.2°F to 98.6°F
10 Pack
25 kDa
10 mm
33 ft.
Supplied in 0.1% sodium azide solution
4°C to 37°C
Ideal for desalting, purification, and molecular separation
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products