Learn More
Spectrum™ Spectra/Por™ Magnetic Weighted Closures
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Brand:  Spectrum™ 132760
 View more versions of this product
Product Code. 11550900
  • 130.00 € / Pack of 2
Explore more special offers
Description
Description
Spectra/Por Magnetic Closure Type:
  • Made from polypropylene with a fluorocarbon coated magnetic SS bar
  • Use on magnetic stir-plate to provide sample stirring for increased dialysis efficiency
  • 4 sealing width sizes: 23mm, 35mm, 55mm and 75mm
  • Autoclavable
  • 10/pkg
  • Use with RC dialysis tubing only
  • Not for use with CE tubing
Specifications
Specifications
Dialysis Tubing Closure
Polypropylene
Autoclavable
Red
0°C to 121°C
2 Pack
Regenerated Cellulose
23 mm
Magnetic
32°F to 250°F
Magnetic fluorocarbon coated stainless steel bar
Use on magnetic stir-plate to provide sample stirring for increased dialysis efficiency, use with RC dialysis tubing only, not for use with CE tubing