Learn More
3M™ Speedglas™ Outer Protection Plate 9100
Product Code. 15809654
Click to view available options
:
Color: Clear
Product Code. 15809654
Brand: 3M™ 7100016691
Only null left
Add to basket
Edge
Add to basket
3M™ Speedglas™ Outer Protection Plate 9100 is designed to protect the filter from spark and spatter.
  • Replacement spare part to the original protection plate if any pits or scratches occur
  • For maximum safety and comfort, make it a habit to check your equipment regularly
  • Regularly clean with a soft cloth and replace if any pits or scratches occur
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Product Type Helmet Outside Protection Plate