Learn More
Spermidine trihydrochloride, 99+%, Thermo Scientific Chemicals
33.47 € - 356.78 €
Chemical Identifiers
CAS | 334-50-9 |
---|---|
Molecular Formula | C7H22Cl3N3 |
Molecular Weight (g/mol) | 254.62 |
MDL Number | MFCD00012918 |
InChI Key | LCNBIHVSOPXFMR-UHFFFAOYSA-N |
Synonym | spermidine trihydrochloride, n-3-aminopropyl-1,4-butanediamine trihydrochloride, spermidine hydrochloride, n1-3-aminopropyl butane-1,4-diamine trihydrochloride, unii-1o14bed398, 1,4-butanediamine, n-3-aminopropyl-, trihydrochloride, spermidine, trihydrochloride, 1,4-butanediamine, n-3-aminopropyl , hydrochloride, n-3-aminopropyl butane-1,4-diamine trihydrochloride, 4-azoniaoctamethylenediammonium trichloride |
PubChem CID | 9539 |
IUPAC Name | N'-(3-aminopropyl)butane-1,4-diamine;trihydrochloride |
SMILES | [H+].[H+].[H+].[Cl-].[Cl-].[Cl-].NCCCCNCCCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10300904
|
Thermo Scientific Acros
215100010 |
1 g | Glass bottle |
33.47 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10430491
|
Thermo Scientific Acros
215100050 |
5 g | Glass bottle |
112.56 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10177463
|
Thermo Scientific Acros
215100250 |
25 g | Glass bottle |
356.78 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
334-50-9 | |
254.62 | |
LCNBIHVSOPXFMR-UHFFFAOYSA-N | |
9539 | |
[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].NCCCCNCCCN |
C7H22Cl3N3 | |
MFCD00012918 | |
spermidine trihydrochloride, n-3-aminopropyl-1,4-butanediamine trihydrochloride, spermidine hydrochloride, n1-3-aminopropyl butane-1,4-diamine trihydrochloride, unii-1o14bed398, 1,4-butanediamine, n-3-aminopropyl-, trihydrochloride, spermidine, trihydrochloride, 1,4-butanediamine, n-3-aminopropyl , hydrochloride, n-3-aminopropyl butane-1,4-diamine trihydrochloride, 4-azoniaoctamethylenediammonium trichloride | |
N'-(3-aminopropyl)butane-1,4-diamine;trihydrochloride |
Specifications
334-50-9 | |
White | |
99% min. (Argentometry) | |
H2N(CH2)4NH(CH2)3NH2·3HCl | |
04, II, 704 | |
spermidine trihydrochloride, n-3-aminopropyl-1,4-butanediamine trihydrochloride, spermidine hydrochloride, n1-3-aminopropyl butane-1,4-diamine trihydrochloride, unii-1o14bed398, 1,4-butanediamine, n-3-aminopropyl-, trihydrochloride, spermidine, trihydrochloride, 1,4-butanediamine, n-3-aminopropyl , hydrochloride, n-3-aminopropyl butane-1,4-diamine trihydrochloride, 4-azoniaoctamethylenediammonium trichloride | |
LCNBIHVSOPXFMR-UHFFFAOYSA-N | |
N'-(3-aminopropyl)butane-1,4-diamine;trihydrochloride | |
9539 | |
99+% | |
Spermidine trihydrochloride, 99+% |
248°C to 252°C | |
Authentic | |
C7H22Cl3N3 | |
MFCD00012918 | |
15, 8869 | |
Solubility in water: soluble | |
[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].NCCCCNCCCN | |
254.62 | |
254.63 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 206-379-0
missing translation for 'rtecsNumber' : EJ7023000
missing translation for 'tsca' : TSCA
- Spermidine trihydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.