missing translation for 'onlineSavingsMsg'
Learn More

Spermine, 97%, Thermo Scientific Chemicals

Product Code. 10488480
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-44-3
Molecular Formula C10H26N4
Molecular Weight (g/mol) 202.35
MDL Number MFCD00008215
InChI Key PFNFFQXMRSDOHW-UHFFFAOYSA-N
Synonym spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl
PubChem CID 1103
ChEBI CHEBI:15746
IUPAC Name N,N'-bis(3-aminopropyl)butane-1,4-diamine
SMILES NCCCNCCCCNCCCN

Specifications

Melting Point 26.0°C to 30.0°C
Density 0.9370g/mL
Boiling Point 130.0°C (0.1 mmHg)
Flash Point 110°C
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2N(CH2)3NH(CH2)4NH(CH2)3NH2
Quantity 5 g
Beilstein 04, II, 704
Merck Index 15, 8870
Specific Gravity 0.937
Formula Weight 202.34
Percent Purity 97%
Chemical Name or Material Spermine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Spermine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'