Learn More
Spermine tetrahydrochloride, 99%, Thermo Scientific Chemicals
Binds to the polyamine modulatory site of NMDA
36.98 € - 464.31 €
Chemical Identifiers
CAS | 306-67-2 |
---|---|
Molecular Formula | C10H30Cl4N4 |
Molecular Weight (g/mol) | 348.18 |
MDL Number | MFCD00012914 |
InChI Key | XLDKUDAXZWHPFH-UHFFFAOYSA-N |
Synonym | spermine tetrahydrochloride, n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride, gerontine tetrahydrochloride, neuridine tetrahydrochloride, n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride, musculamine tetrahydrochloride, geontine tetrahydrochloride, spermine hcl, 1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride, spermine, tetrahydrochloride |
PubChem CID | 9384 |
IUPAC Name | N,N'-bis(3-aminopropyl)butane-1,4-diamine;tetrahydrochloride |
SMILES | [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15445329
|
Thermo Scientific Alfa Aesar
J63060.03 |
1 g |
36.98 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15455329
|
Thermo Scientific Alfa Aesar
J63060.06 |
5 g |
119.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15465329
|
Thermo Scientific Alfa Aesar
J63060.14 |
25 g |
464.31 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Spermine tetrahydrochloride, is used to precipitate DNA from low salt aqueous buffers. Spermine tetrahydrochloride was used to precipitate DNA for flow cytometric analysis from fresh frozen breast cancer tissue, archival tissue of salivary gland, renal and thyroid tumors2 and red blood cells of chicken.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSpermine tetrahydrochloride, is used to precipitate DNA from low salt aqueous buffers. Spermine tetrahydrochloride was used to precipitate DNA for flow cytometric analysis from fresh frozen breast cancer tissue, archival tissue of salivary gland, renal and thyroid tumors2 and red blood cells of chicken.
Solubility
Soluble in water (348.2 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Store under inert gas. Keep away from strong oxidizing agents.
Chemical Identifiers
306-67-2 | |
348.18 | |
XLDKUDAXZWHPFH-UHFFFAOYSA-N | |
9384 | |
[H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN |
C10H30Cl4N4 | |
MFCD00012914 | |
spermine tetrahydrochloride, n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride, gerontine tetrahydrochloride, neuridine tetrahydrochloride, n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride, musculamine tetrahydrochloride, geontine tetrahydrochloride, spermine hcl, 1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride, spermine, tetrahydrochloride | |
N,N'-bis(3-aminopropyl)butane-1,4-diamine;tetrahydrochloride |
Specifications
306-67-2 | |
C10H30Cl4N4 | |
3911771 | |
Soluble in water (348.2g/L at 20°C). | |
[H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN | |
348.18 | |
348.19 | |
Powder |
White | |
MFCD00012914 | |
spermine tetrahydrochloride, n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride, gerontine tetrahydrochloride, neuridine tetrahydrochloride, n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride, musculamine tetrahydrochloride, geontine tetrahydrochloride, spermine hcl, 1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride, spermine, tetrahydrochloride | |
XLDKUDAXZWHPFH-UHFFFAOYSA-N | |
N,N'-bis(3-aminopropyl)butane-1,4-diamine;tetrahydrochloride | |
9384 | |
99% | |
Spermine tetrahydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 206-189-8
missing translation for 'rtecsNumber' : EJ7230000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.