missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPEX CertiPrep™ Accessories for Pipet Washer and Dryer
Accessories for use with Pipet Washer and Dryer
Brand: SPEX CertiPrep™ PIPPUMP-115V
This item is not returnable.
View return policy
Description
- Pipette Washer Pump, 230V
- For use with SPEX CertiPrep Pipette Washer/Dryer.
- Durable polyethylene construction
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Specifications
For pipet washer and dryer | |
Pipette Washer Pump, 115V |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction