Learn More
SPEX CertiPrep™ Epoxy-coated steel Vial Rack
Use to hold vials for storage and handling.
Brand:  SPEX CertiPrep™ 6805
Product Code. 15320492
  • 178.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Durable epoxy-coated steel construction.
  • Holds up to six of the following large grinding vials:6801 Largegrinding vial set 6871 large CR-free grinding vial set
  • Included with the purchase of each 6870L Large Grinding Vial Accessory Package
Specifications
Specifications
Vial Rack
Holds large grinding vials for storage and handling
Freezer/Mill™ Cryogenic Grinder