Learn More
Sphingomyelin, MedChemExpress
MedChemExpress Sphingomyelin is a eukaryotic sphingolipid and one of the major constituents of cell membranes and particularly abundant in the myelin sheath that surrounds neuronal axons. Sphingomyelin plays an important role in cell processes, the regulation of inflammatory responses, and signal transduction. Sphingomyelin metabolism is associated with various central nervous system diseases and Niemann-Pick disease.
Brand:  Medchem Express HY-113498
Encompass_Preferred
 View more versions of this product
Product Code. 16494778
  • 70.20 € / 10mg
Explore more special offers
Description
Description
Sphingomyelin is a eukaryotic sphingolipid and one of the major constituents of cell membranes and particularly abundant in the myelin sheath that surrounds neuronal axons. Sphingomyelin plays an important role in cell processes, the regulation of inflammatory responses, and signal transduction. Sphingomyelin metabolism is associated with various central nervous system diseases and Niemann-Pick disease.
Specifications
Specifications
Sphingomyelin
White
10 mg
[Sphingomyelin]
732.08
Research
85187-10-6
Research
Ethanol : 25 mg/mL (34.15 mM; Need ultrasonic)
732.08
95.0%
Solid