missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPI1 Mouse anti-Human, Clone: 2H3D3, Proteintech
Mouse Monoclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | SPI1 |
---|---|
Clone | 2H3D3 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828665
|
Proteintech
66618-2-IG-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16818665
|
Proteintech
66618-2-IG-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes an ETS-domain transcription factor that activates gene expression during myeloid and B-lymphoid cell development. The nuclear protein binds to a purine-rich sequence known as the PU-box found near the promoters of target genes, and regulates their expression in coordination with other transcription factors and cofactors. The protein can also regulate alternative splicing of target genes. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
SPI1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
31 kDa transforming protein, OF, PU.1, SFPI1, SPI 1, SPI A, SPI1, Transcription factor PU.1 | |
SPI1 | |
IgG1 | |
Protein G | |
Antibody |
2H3D3 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P17947 | |
6688 | |
Peptide | |
Primary | |
-20°C | |
SPI1 |