Learn More
Heidolph™ Spider with Flasks
For use with rotary evaporators
Brand:  Heidolph™ 15-300-005-07
Product Code. 10025851
  • 841.00 € / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Comes with a standard NS 29/32 joint size but are also available with NS 24/29 ground joints
Specifications
Specifications
Glass
Rotary evaporators
100 mL (per flask)
Spider with Flasks
5 flasks (100 mL)