missing translation for 'onlineSavingsMsg'
Learn More

Spike Sample Analysis Standard I For CLP, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

344.00 € - 600.00 €

Specifications

Chemical Name or Material ICP Spike Sample 1
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition (by Analyte or Components) Components in 5% HNO3/tr Tartaric Acid/tr HF:
Al    •200μg/mL
Sb    •50μg/mL
As    •200μg/mL
Ba    •200μg/mL
Be    •5μg/mL
Cd    •5μg/mL
Cr    •20μg/mL
Co    •50μg/mL
Cu    •25μg/mL
Fe    •100μg/mL
Pb    •50μg/mL
Mn    •50μg/mL
Ni    •50μg/mL
Se    •200μg/mL
Ag    •5μg/mL
Tl    •200μg/mL
V    •50μg/mL
Zn    •50μg/mL
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11465968
View Documents
SPEX Certiprep
SPIKE-1
125 mL Triple-leached LDPE bottle
344.00 €
125mL
Please to purchase this item. Need a web account? Register with us today!
11475968
View Documents
SPEX Certiprep
SPIKE-1-500
500 mL Triple-leached LDPE bottle
600.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Al200μg/mL
As200μg/mL
Ba200μg/mL
Se200μg/mL
Tl200μg/mL
Fe100μg/mL
Co50μg/mL
Mn50μg/mL
Ni50μg/mL
Pb50μg/mL
Sb50μg/mL
V50μg/mL
Zn50μg/mL
Cu25μg/mL
Cr20μg/mL
Ag5μg/mL
Be5μg/mL
Cd5μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

ICP Spike Sample 1
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Components in 5% HNO3/tr Tartaric Acid/tr HF:
Al    •200μg/mL
Sb    •50μg/mL
As    •200μg/mL
Ba    •200μg/mL
Be    •5μg/mL
Cd    •5μg/mL
Cr    •20μg/mL
Co    •50μg/mL
Cu    •25μg/mL
Fe    •100μg/mL
Pb    •50μg/mL
Mn    •50μg/mL
Ni    •50μg/mL
Se    •200μg/mL
Ag    •5μg/mL
Tl    •200μg/mL
V    •50μg/mL
Zn    •50μg/mL
Triple-leached LDPE bottle
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
5% HNO3/tr Tart. Acid/tr HF
Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

missing translation for 'dotInformation' : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Product Certifications

Product intended for laboratory use only