missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Spill tray, orange painted metal, 40L capacity
Brand: Manutan A016814

This item is not returnable.
View return policy
Description
Suitable for the storage and handling of containers that may leak. Trays nest for space-saving storage. Thick sheet steel with lacquered orange paint finish.- Capacity: 40L
- With handles
- Dimensions (L x W x H): 700mm x 450mm x 140mm
- Weight: 8kg
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction