missing translation for 'onlineSavingsMsg'
Learn More

Cole Parmer Spill-X Refill

Cole Parmer Spill-X Refill is an easy-to-store carton of six handy polypropylene containers to replace specific or multipurpose Spill Treatment Kit containers.

62705.00 €

Specifications

Material Polypropylene
Type Spill Treatment Refill
For Use With (Equipment) Spill Treatment Kit Containers
Model Spill-X Refill
Packaging Quantity 6/Cs.
View More Specs

Products 1
Product Code Brand For Use With (Application) Color Price Quantity & Availability  
Product Code Brand For Use With (Application) Color Price Quantity & Availability  
13141176
View Documents
Cole Parmer
86292-50
Acid Agent Spill Red
62705.00 €
Pack of 6
Estimated Shipment: 25-02-2025
to see stock.
Add to basket
 
Description

Description

Cole Parmer Spill-X Refill is an easy-to-store carton of six handy polypropylene containers to replace specific or multipurpose Spill Treatment Kit containers.

Specifications

Specifications

Polypropylene
Spill Treatment Kit Containers
6/Cs.
Spill Treatment Refill
Spill-X Refill
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • X6 SPILL-X-A ACID AGENT REFILL,

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : magnesium oxide, fullers earth, sodium carbonate

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.