Learn More
Corning™ Spin-X™ UF Concentrators
Disposable, single use only ultrafiltration devices with polyethersulfone membranes (PES) for the centrifugal concentration and/or purification of biological fluids and aqueous solutions
137.95 € - 207.29 €
Specifications
Cap Material | Polypropylene |
---|---|
Color | White |
Filter Type | Centrifuge Filter |
Material | PES |
Material (Membrane) | PES |
Product Code | Brand | Volume (Metric) | Quantity | Filtration Area | MWCO | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Quantity | Filtration Area | MWCO | Price | Quantity & Availability | ||
11383785
|
Corning™
431477 |
500 μL | 25/Pk. | 0.5 cm2 | 5000 |
137.95 €
Pack of 25 |
Estimated Shipment: 08-04-2025
Log in to see stock. |
||
11303795
|
Corning™
431479 |
500 μL | 25/Pk. | 0.5 cm2 | 30 kDa |
139.99 €
Pack of 25 |
Estimated Shipment: 08-04-2025
Log in to see stock. |
||
11393785
|
Corning™
431478 |
500 μL | 25/Pk. | 0.5 cm2 | 10 kDa |
139.99 €
Pack of 25 |
In Stock
Log in to see stock. |
||
11313795
|
Corning™
431480 |
500 μL | 25/Pk. | 0.5 cm2 | 50,000 |
139.99 €
Pack of 25 |
In Stock
Log in to see stock. |
||
11373795
|
Corning™
431486 |
6 mL | 25/Pk. | 2.5 cm2 | 100,000 |
207.29 €
Pack of 25 |
In Stock
Log in to see stock. |
||
11333795
|
Corning™
431482 |
6 mL | 25/Pk. | 2.5 cm2 | 5000 |
207.29 €
Pack of 25 |
In Stock
Log in to see stock. |
||
11343795
|
Corning™
431483 |
6 mL | 25/Pk. | 2.5 cm2 | 10 kDa |
207.29 €
Pack of 25 |
Estimated Shipment: 08-04-2025
Log in to see stock. |
||
11323795
|
Corning™
431481 |
500 μL | 25/Pk. | 0.5 cm2 | 100,000 |
139.99 €
Pack of 25 |
In Stock
Log in to see stock. |
||
11363795
|
Corning™
431485 |
6 mL | 25/Pk. | 2.5 cm2 | 50,000 |
207.29 €
Pack of 25 |
In Stock
Log in to see stock. |
||
11353795
|
Corning™
431484 |
6 mL | 25/Pk. | 2.5 cm2 | 30 kDa |
207.29 €
Pack of 25 |
Estimated Shipment: 08-04-2025
Log in to see stock. |
||
11383795
|
Corning™
431487 |
20 mL | 12/Pk. | 6 cm2 | 5000 |
148.72 €
Pack of 12 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
11393795
|
Corning™
431488 |
20 mL | 12/Pk. | 6 cm2 | 10 kDa |
150.00 €
Pack of 12 |
In Stock
Log in to see stock. |
||
11303805
|
Corning™
431489 |
20 mL | 12/Pk. | 6 cm2 | 30 kDa |
148.72 €
Pack of 12 |
Estimated Shipment: 08-04-2025
Log in to see stock. |
||
11323805
|
Corning™
431491 |
20 mL | 12/Pk. | 6 cm2 | 100,000 |
148.72 €
Pack of 12 |
Estimated Shipment: 08-04-2025
Log in to see stock. |
||
11313805
|
Corning™
431490 |
20 mL | 12/Pk. | 6 cm2 | 50,000 |
148.72 €
Pack of 12 |
Estimated Shipment: 08-04-2025
Log in to see stock. |
||
Description
Spin-X UF centrifugal concentrators offer a simple, one step procedure for concentrating or desalting proteins and other biomolecules with 90% or better recovery.
- The vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides fast, high speed concentrating, even with particle laden solutions.
- Low binding polyethersulfone (PES) membranes are available with five molecular weight cut-offs (MWCO): 5,000, 10,000, 30,000, 50,000, and 100,000 to meet all of your concentrating needs. Choose an MWCO half to a third smaller than the protein to be concentrated.
- The MWCO and graduations are printed on the side of the concentrator tube to avoid mix-ups.
- Spin-X UF 6 and 20 concentrators can be used with either swinging bucket or fixed angle rotors. Spin-X UF 500 concentrators require fixed angle rotors.
Specifications
Polypropylene | |
Centrifuge Filter | |
PES | |
Ultrafiltration Concentrator |
White | |
PES | |
Concentration, desalting of proteins, enzymes, monoclonal antibodies, immunoglobulins, removal of labeled amino acids and nucleotides, HPLC sample preparation, deproteinization of samples, recovery of biomolecules from cell culture supernatants, lysates |