missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SPINeasy™ DNA Purification Kit

Designed for fast and efficient cleanup of pre-purified DNA samples. Removes a wide range of inhibitory compounds, such as those found in environmental samples (humic acid, heavy metals, etc.), as well as organic compounds that may co-purify with DNA from other extraction procedures.

Brand:  MP Biomedicals™ 116548050


Product Code. 17864126

  • 268.00 € / 50 preps

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • The SPINeasy DNA Purification Kit is designed for the removal of contaminants that may co-purify with DNA and inhibit downstream enzymatic, PCR applications.
  • Inhibitory compounds such as those found in environmental samples (humic acid, heavy metals, etc.), as well as organic compounds that may co-purify with DNA from other extraction procedures, are efficiently and quickly removed using this kit.
  • The kit offers two choices of binding buffers. Buffer P1 is recommended for purification of samples with slight contamination of humic acids (light brownish color) or other PCR inhibitors. Buffer P1HA contains a novel and proprietary humic acid removal technology, which is recommended for processing samples contaminated with a considerable amount of humic substances (strong brownish color).
  • The kit has been validated with genomic DNA isolated from various problematic soil samples, as well as with DNA samples spiked with humic acid molecules. The data shows that the kit can clean up to 50 µg DNA with a size ranging from 200 bp to high molecular weight, with expected recovery of > 80%.
  • Cleaned-up DNA samples can be readily used in downstream applications, including long fragment PCR, qPCR, and next-generation sequencing (16S and whole genome).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at Room Temperature, Dry Environment
25 to 200 µL DNA preparations > 200 bp
50 Preps
Spin Column
Buffer P1 50 mL, Buffer P1HA 50mL, Buffer P2 40 mL, Buffer P3 40 mL, Buffer P4 6 mL, 50x Column P, 100 x 2.0 mL collection tubes, 50 x 1.5 mL collection tubes
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • SPINeasy DNA Purification Kit

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Guanidine Hydrochloride

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.