missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Spinner Direct Top Drive Motor
High torque, low rpm stirrer designed to maintain constant low speed.
Brand: Corning™ 402645
This item is not returnable.
View return policy
Description
- High torque, low rpm stirrer designed to maintain constant low speed.
- Gearhead stirrer delivers 14.5 in lbs of torque.
- Maximum speed is 350 rpm.
- Weight of motor is 9 lbs (4.1kg).
- Available with 120VAC 60 Hz or 230VAC 50Hz.
Order Info
ForUseWithEquipment

Specifications
Proculture Spinner Flasks | |
Non-sterile |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Spinner Direct Top Drive Motor > For use with Proculture Spinner Flasks
Spot an opportunity for improvement?Share a Content Correction