Learn More
Novus Biologicals™ Spleen Tissue Slides (Normal)- Paraffin
Brand:  Novus Biologicals™ NBP2-30202
Product Code. 18759083
  • 291.00 € / Pack of 5
Explore more special offers
Description
Description
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
Specifications
Specifications
Human
Immunohistochemistry, Immunohistochemistry-Paraffin, In-situ Hybridization, Nucleic Acid Extraction
Human
Spleen Tissue Slides (Normal)
This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Normal