missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ SPME Arrow Liner
216.94 € - 349.78 €
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | |||||
15840705
|
Thermo Scientific™
453A0411 |
SPME Arrow Liner ID 1.3 mm | 3 Pk. |
349.78 €
Pack of 3 |
Estimated Shipment: 02-05-2025
Log in to see stock. |
|||||
15850705
|
Thermo Scientific™
453A0415 |
SPME Arrow Liner ID 1.7 mm x 6.5 mm x 78.5 mm | 3 Pk |
349.78 €
Pack of 3 |
Estimated Shipment: 17-04-2025
Log in to see stock. |
|||||
17644145
|
Thermo Scientific™
453A0415-UI |
SPME Arrow Liner ID 1.7 mm x 6.5 mm x 78.5 mm | 5 Pk. |
216.94 €
Pack of 5 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
|||||
Description
Specifications
SPME Arrow |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ SPME Arrow Liner