missing translation for 'onlineSavingsMsg'
Learn More

Neogen Sponge-Stick

Product Code. 17293744
Encompass
Click to view available options
Includes:
10 mL Letheen Broth
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 17293744

Brand: Neogen 7100010357

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Uses simplified packaging to minimize handling and preparation, and reduces waste.

  • Stick-mounted sponge design allows the ability to access hard to reach areas without directly handling the sponge.
  • Stick-mounted sponge design provides the ability to easily snap off the sponge from the handle after swabbing.
  • Simplified packaging including Sponge-Stick, sample bag and gloves, minimizes handling and preparation and reduces waste.
  • 1.5 x 3 in. biocide-free sponge does not affect organism viability.
  • 6 x 11 in. sample bag with a 30 oz./0.89 L capacity included for safe, convenient transportation to lab.

Specifications

Product Type Sponge-Stick
Length (English) 3 in.
Length (Metric) 7.6 cm
Width (English) 1.5 in.
Width (Metric) 3.8 cm
Packaging Quantity 100/Cs.
Includes 10 mL Letheen Broth
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.