Learn More
Tocris Bioscience™ SPP 86
Potent RET inhibitor
Brand:  Tocris Bioscience™ 5391/10
176.24 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18770498
  • 224.00 € / 10mg
Explore more special offers
Description
Description
SPP 86 is a potent RET inhibitor (IC50 = 8 nM). Also exhibits inhibitory activity at EphA1, FGFR1, Flt4, Lck and Yes. Inhibits proliferation of HCT 116 and TPC1 cells in vitro. Cell permeable.
Specifications
Specifications
>98%
277.32
10 mg
SPP 86
SPP 86